Hexestrol
Hexestrol
Catalog # EHERC14202800
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC[C@H]([C@H](CC)c1ccc(O)cc1)c2ccc(O)cc2
- Inchi: InChI=1S/C18H22O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h5-12,17-20H,3-4H2,1-2H3/t17-,18+
- IUPAC: 4-[(3S,4R)-4-(4-hydroxyphenyl)hexan-3-yl]phenol
Specifications
Specifications
- Description:Hexestrol
- Cat. No.:EHERC14202800
- Formula:C₁₈H₂₂O₂
- Storage conditions:20 °C
- Hazard class description:GHS08
- GHS signal:Danger
- Synonym(s):Phenol, 4,4'-(1,2-diethyl-1,2-ethanediyl)bis-, (R*,S*)-; Phenol, 4,4'-(1,2-diethylethylene)di-, meso- (8CI); Cycloestrol; Erythrohexestrol; Estrifar; Estronal; Extra-Plex; Hexanoestrol; Hexestrofen; Hexestrol; Hexoestrol; Hexron; Hormoestrol; Mesohexestrol; NSC 9894; Sinestrol; Synestrol; Synoestrol; Synthovo; Syntrogene; meso-3,4-Bis(p-hydroxyphenyl)-n-hexane; meso-3,4-Di(p-hydroxyphenyl)-n-hexane; meso-Hexestrol; Phenol, 4,4'-[(1R,2S)-1,2-diethyl-1,2-ethanediyl]bis-, rel-
- Molecular weight:270,37
- CAS No.:84-16-2
- Shipping conditions:RT
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



