OptiMATE Lower Fluidics
OptiMATE lower fluidics
Catalog # BECMD12219
New Product
Supplier: Beckman Coulter
Jump to:
Product Details & Documents
The OptiMATE Lower Fluidics includes a cap with a dip tube and vent tubing with a filter. This cap is used to connect general reagent bottles to the upper fluidics path. Contains 4 units. Endotoxin: ≤10 EU/device.
Specifications
- Description:OptiMATE lower fluidics