Ampicillin ≥95%
Bottle
Catalog # APOSBUP19184-1ML
New Product
Supplier: Apollo Scientific
Ampicillin ≥95%
Catalog # APOSBUP19184-1ML
Supplier: Apollo Scientific
New Product
CAS Number: 69-53-4
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
- Store at 2 to 8 °C
- CAS Number:69-53-4
- Formula:C₁₆H₁₉N₃O₄S
- Molecular Weight:349,4 g/mol
- Storage Temperature:Refrigerator
- EINECS:200-709-7
Specifications
Specifications
- Pack type:Bottle



