Order Entry
Germany
Orders LinkContactUsLinkComponent
Pyrene 98%

Pyrene 98%

Bottle
Catalog # APOSOR72328-500G
Toxic
Environment
Supplier:   Apollo Scientific
Pyrene 98%
Catalog # APOSOR72328-500G
Supplier:  Apollo Scientific
CAS Number:  129-00-0
Pyrene 98%

Bottle

CAS Number:  129-00-0
Price
81,10 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:129-00-0
  • Formula:C₁₆H₁₀
  • MDL Number:MFCD00004136
  • Molecular Weight:202,25059 g/mol
  • UN:3077
Specifications

Specifications

  • Pack type:
    Bottle

Specifications