Order Entry
Germany
Orders LinkContactUsLinkComponent
Ectoine 98%

Ectoine 98%

Irritant
Supplier:   Apollo Scientific
Ectoine 98%
Supplier:  Apollo Scientific
Certificates

5 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  (S)-2-Methyl-3,4,5,6-tetrahydropyrimidine-4-carboxylic acid
  • CAS Number:96702-03-3
  • Formula:C₆H₁₀N₂O₂
  • MDL Number:MFCD03419286
  • Molecular Weight:142,157 g/mol
  • EINECS:431-910-1
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 82,50 €
    82,50 €
    1 * 25 g
       82,50 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 273,00 €
    273,00 €
    1 * 100 g
       273,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 734,00 €
    734,00 €
    1 * 500 g
       734,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.