Methyl red indicator
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/reagent.
- Non-hazardous
Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
- CAS Number:493-52-7
- Formula:C₁₅H₁₅N₃O₂
- MDL Number:MFCD00002425
- Molecular Weight:269,3 g/mol
- EINECS:207-776-1
5 Options Available Below
Product Details & Documents
Usage: Organic building block/reagent.
- Non-hazardous
Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
- CAS Number:493-52-7
- Formula:C₁₅H₁₅N₃O₂
- MDL Number:MFCD00002425
- Molecular Weight:269,3 g/mol
- EINECS:207-776-1
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



