Order Entry
Germany
Orders LinkContactUsLinkComponent
Neutral red ≥98%

Neutral red ≥98%

Bottle
Catalog # APOSBUP18189-500MG
New Product
Supplier:   Apollo Scientific
Neutral red ≥98%
Catalog # APOSBUP18189-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  553-24-2
Neutral red ≥98%

Bottle

CAS Number:  553-24-2
Price
52,10 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
  • CAS Number:553-24-2
  • Formula:C₁₅H₁₇ClN₄
  • Molecular Weight:288,77 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.