Order Entry
Germany
Orders LinkContactUsLinkComponent
Thioisonicotinamide ≥98%

Thioisonicotinamide ≥98%

Bottle
Catalog # APOSBUP17646-200MG
New Product
Supplier:   Apollo Scientific
Thioisonicotinamide ≥98%
Catalog # APOSBUP17646-200MG
Supplier:  Apollo Scientific
New Product
CAS Number:  2196-13-6
Thioisonicotinamide ≥98%

Bottle

CAS Number:  2196-13-6
Price
37,70 €
Log in for availability
1 * 200 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  Pyridine-4-carbothioamide
  • CAS Number:2196-13-6
  • Formula:C₆H₆N₂S
  • Molecular Weight:138,19 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications