Ectoine ≥95%
Bottle
Catalog # APOSBUP17528-100MG
New Product
Supplier: Apollo Scientific
Ectoine ≥95%
Catalog # APOSBUP17528-100MG
Supplier: Apollo Scientific
New Product
CAS Number: 96702-03-3
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms: (S)-2-Methyl-3,4,5,6-tetrahydropyrimidine-4-carboxylic acid
- CAS Number:96702-03-3
- Formula:C₆H₁₀N₂O₂
- Molecular Weight:142,16 g/mol
Specifications
Specifications
- Pack type:Bottle
- Cat. No.:APOSBUP17528-100MG



