Thymolphthalein indicator
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/ reagent.
Synonyms: 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
- CAS Number:125-20-2
- Formula:C₂₈H₃₀O₄
- MDL Number:MFCD00005909
- Molecular Weight:430,54 g/mol
4 Options Available Below
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyms: 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
- CAS Number:125-20-2
- Formula:C₂₈H₃₀O₄
- MDL Number:MFCD00005909
- Molecular Weight:430,54 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



