Order Entry
Germany
Orders LinkContactUsLinkComponent
Ninhydrin ≥98%

Ninhydrin ≥98%

Bottle
Catalog # APOSBUP19645-1G
New Product
Supplier:   Apollo Scientific
Ninhydrin ≥98%
Catalog # APOSBUP19645-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  485-47-2
Ninhydrin ≥98%

Bottle

CAS Number:  485-47-2
Price
37,70 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
  • CAS Number:485-47-2
  • Formula:C₉H₆O₄
  • Molecular Weight:178,14 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications