Order Entry
Germany
Orders LinkContactUsLinkComponent
Serving Trolley, VC

Serving Trolley, VC

Serving trolley, 800×600 mm, total capacity: 300 kg, 18 kg, 3
Catalog # HUPF71020-1-VC
New Product
Supplier:   HUPFER METALLWERKE
Serving Trolley, VC
Catalog # HUPF71020-1-VC
Supplier:  HUPFER METALLWERKE
New Product
Serving Trolley, VC

Serving trolley, 800×600 mm, total capacity: 300 kg, 18 kg, 3

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
  • Wheel details: Plastic

    Equipped with: Buffers, fixable castors

    Stainless steel

Specifications
  • Description:
    Serving trolley, 800×600 mm, total capacity: 300 kg
  • Shelves:
    3
  • Weight:
    18 kg
  • Cat. No.:
    HUPF71020-1-VC
  • Length:
    1020 mm
  • Wheel Ø:
    125 mm
  • Model:
    VC 8×6/3
  • Material:
    Stainless steel
  • Height:
    1105 mm
  • Wheel type:
    4 swivel castors
  • Depth:
    685 mm