Orders LinkContactUsLinkComponent
Handheld Digital Microscope Edge AF4935ZTLE
Handheld Digital Microscope Edge AF4935ZTLE
Handheld digital microscope, 114H×34D mm, AF4935ZTLE, 1.3 Megapixel (1280×960)
Catalog # IDCPAF4935ZTLE
New Product
Handheld Digital Microscope Edge AF4935ZTLE
Catalog # IDCPAF4935ZTLE
New Product
Handheld Digital Microscope Edge AF4935ZTLE
Handheld digital microscope, 114H×34D mm, AF4935ZTLE, 1.3 Megapixel (1280×960)
Price
805,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The AF4935ZTLE is a feature-packed long working distance microscope with a versatile magnification range of 10x ~ 200x. AF4935ZTLE is equipped with a polarizer, and features FLC, AMR, EDOF, and EDR, making it ideal for a wide range of tasks. This model is also compatible with the WF-20 Wi-Fi streamer for wireless connectivity.
​1.3 Megapixel: The advanced CMOS image sensor can transmit fluid and crisp images at 15 fps with resolution up to 1280×960
​Long Working Distance (LWD) Optics: The LWD optics yield more working space between the object and the microscope, making it ideal for applications such as repair or assembly.
​Flexible LED Control (FLC): Tasking with software, the FLC provides a six-level intensity adjustment capability for active LED quadrants.
​Adjustable Polarizer: The built-in adjustable polarizer removes unwanted reflection or glare from the object's surface for better contrast.
Extended Depth of Field (EDOF): Viewing rough surfaces with height ranges out of depth of focus, the EDOF can take several images at different focuses and stack them automatically within a click.
Extended Dynamic Range (EDR): Observing high contrast or reflective surface, the EDR can help to reveal the details of dark or bright areas by stacking images taken at different exposure levels.
Auto and Manual Refocusing: Using mouse inputs, the refocusing capability helps fine-tune the focus, either automatically by clicking at the point of interest or manually by scrolling the mouse.
Automatic Magnification Readout (AMR): The Automatic Magnification Readout (AMR) detects the magnification in use, eliminating manual input errors and enhancing measurement accuracy in tasks with frequent magnification adjustments.
​Adaptable Interface: The adaptable interface provides the capability of transforming Dino-Lite into a wireless microscope when attaching with the WF-20.
Specifications
  • Magnification:
    Adjustable, 10 ~ 200x
  • Model:
    AF4935ZTLE
  • Resolution:
    1.3 Megapixel (1280×960)
  • Description:
    Handheld digital microscope, 114H×34D mm
  • Cat. No.:
    IDCPAF4935ZTLE
  • Illumination:
    White light, 8 LEDs
  • Frame rate:
    15 fps at 1.3 MP, MJPEG; 30 fps at VGA, MJPEG
  • Software:
    DinoCapture 2.0, DinoCapture 3.0, DinoXcope
  • Weight:
    Approx. 110 g, incl. cable and cap (N3C-O)