Compressed Air System
Compressed air system CP-MP, it provides pressure support during cooling and dispensing phases; This accessory is required to enable controlled external dispensing through the external dispensing station DW-MP
Catalog # ESPICP-MP
New Product
Supplier: RAYPA
Jump to:
Product Details & Documents
It provides pressure support during cooling and dispensing phases. This accessory is required to enable controlled external dispensing through the external dispensing station DW-MP.
: Factory installation required. The AE-60-MP, AE-80-MP, and AE-100-MP models include this accessory by default.
Specifications
- Description:Compressed air system CP-MP, it provides pressure support during cooling and dispensing phases; This accessory is required to enable controlled external dispensing through the external dispensing station DW-MP