Orders LinkContactUsLinkComponent
Embedded Thermal Printer
Embedded Thermal Printer
Embedded thermal printer for media preparators IT/MP, allows physical documentation of the sterilisation cycle results; 85×85 mm (L×H); 8 dots/mm print quality, 203 dpi resolution
Catalog # ESPIIT/MP
New Product
Supplier:   RAYPA
Embedded Thermal Printer
Catalog # ESPIIT/MP
Supplier:  RAYPA
New Product
Embedded Thermal Printer
Embedded Thermal Printer
Embedded thermal printer for media preparators IT/MP, allows physical documentation of the sterilisation cycle results; 85×85 mm (L×H); 8 dots/mm print quality, 203 dpi resolution
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Allows quick printing on paper of the most relevant results from each sterilisation cycle carried out in the media preparators.
  • Integrated location in the front control panel of the autoclave
  • Print settings and ticket retrieval managed from the autoclave screen
  • Selectable print cadence: from 10 to 240 s
  • Prints key cycle data: program name, temperature evolution, date, time, and error messages
  • Paper consumable can be replaced by the user
  • Energy-saving mode
The external dot matrix printer provides physical documentation of each sterilisation cycle, printing program and cycle numbers, temperature, pressure, date, time, and error messages. Two printing modes are selectable from the autoclave screen: simplified printing at the end of the cycle or extended printing during the cycle with detailed parameter tracking.
Recommended for situations where PC software use is not feasible and for improving traceability of experiments or production processes.
Achtung! Bitte beachten: Factory installation required.
Specifications
  • Description:
    Embedded thermal printer for media preparators IT/MP, allows physical documentation of the sterilisation cycle results; 85×85 mm (L×H); 8 dots/mm print quality, 203 dpi resolution