Orders LinkContactUsLinkComponent
Barcode Scanner
Barcode Scanner
USB barcode scanner for Reading individual labels for each processed load and identifying each batch, Reading individual labels for each processed load and identifying each batch
Catalog # ESPIBAR-SCAN
Supplier:   RAYPA
Barcode Scanner
Catalog # ESPIBAR-SCAN
Supplier:  RAYPA
Barcode Scanner
Barcode Scanner
USB barcode scanner for Reading individual labels for each processed load and identifying each batch, Reading individual labels for each processed load and identifying each batch
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Allows reading individual labels for each processed load and identifying each batch.
  • The purchase of this accessory includes: 1 barcode scanner, 1 connection cable and 1 scanner holder
  • Connection: USB
All top line autoclaves include RAYPAtrace, a simple and professional solution for the documentation and complete traceability of processed batches. To optimise the process, it is recommended to use a label printer (Ref. ITS-LAB) and a barcode scanner (Ref. BAR-SCAN), tools that facilitate the identification and tracking of each batch through individual labels.
Specifications
  • Description:
    USB barcode scanner for Reading individual labels for each processed load and identifying each batch
  • For:
    Reading individual labels for each processed load and identifying each batch