Order Entry
Germany
Orders LinkContactUsLinkComponent
Vancomycin hydrochloride

Vancomycin hydrochloride

Supplier:   G-Biosciences
Vancomycin hydrochloride
Supplier:  G-Biosciences
Certificates
Vancomycin hydrochloride

3 Options Available Below

Product Details & Documents

Vancomycin is a glycopeptide antibiotic that exerts its action by preventing the transfer and addition of the muramylpentapeptides that comprise the peptidoglycan. Vancomycin is often used synergistically with cefotaxime or carbenicillin against Agrobacterium.

  • Molecular Biology Grade
  • Tested against both sensitive and resistant cells
  • Potency of greater than 1000 mcg/mg
Synonyms:  (3S,6R,7R,22R,23S,26S,36R,38aR)-3-(2-Amino-2-oxoethyl)-44-[[2-O-(3-amino-2,3,6-trideoxy-3-C-methyl-α-L-lyxo-hexopyranosyl)-β-D-glucopyranosyl]oxy]-10,19-dichloro-2,3,4,5,6,7,23,24,25,26,36,37,38,38a-tetradecahydro-7,22,28,30,32-pentahydroxy-6-[[(2R)-4-methyl-2-(methylamino)-1-oxopentyl]amino]-2,5,24,38,39-pentaoxo-22H-8,11:18,21-Dietheno-23,36-(iminomethano)-13,16:31,35-dimetheno-1H,16H-[1,6,9]oxadiazacyclohexadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine-26-carboxylic acid xhydrochloride, Vancomycin supplement
  • CAS Number:1404-93-9
  • Formula:C₆₆H₇₅Cl₂N₉O₂₄·HCl
  • Molecular Weight:1485,71 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Vial
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 137,00 €
    137,00 €
    1 * 1 g
       137,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 308,00 €
    308,00 €
    1 * 5 g
       308,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 671,00 €
    671,00 €
    1 * 25 g
       671,00 €