Orders LinkContactUsLinkComponent
b.safe Level Control Set
b.safe Level Control Set
Level control set, 1-channel; No. of containers to be monitored: 1
Catalog # BOHLR9950-05
New Product
Supplier:   Bohlender
b.safe Level Control Set
Catalog # BOHLR9950-05
Supplier:  Bohlender
New Product
b.safe Level Control Set
b.safe Level Control Set
Level control set, 1-channel; No. of containers to be monitored: 1
Price
412,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Monitoring of the fill level or empty level of containers and canisters made of non-conductive materials like plastics or glass. Avoid spill over of collection canisters or storage canisters running empty by timely alarms.
Set for monitoring the fill level or empty level of containers and canisters made of non-conductive materials consisting of b.safe Disk Sensor, signal cable and b.safe Mini-Display Table. The Disk Sensor can be set to being an indicator for fill level or empty level as required, please observe the operating instructions.
The sensor is simply fixed on the canister using the included hook-and-loop tape or the self-adhesive hook-and-loop strip. The sensor is connected to the alarm box b.safe Mini Display with the signal cable. The compact plastic housing can be set up in the workspace. Once fill level to be monitored is reached, the status LED on the Mini-Display lights up red and an acoustic alarm starts.
The acoustic alarm can be paused for 5 minutes at a time. As soon as the canister has been exchanged, the Mini-Display can be reset and the control lamp turns to green. b.safe Mini-Display Table including power supply (Input: AC 100 to 240 V/50-60 Hz, 1 A; Output: 24 V DC, 0,5 A) with plugs for EU, Australia, USA, Japan and UK. Not suitable for use in EX zones or with an isolating amplifier.
Delivery information: Supplied with 1×Mini-Display Table, 1 channel 1×Disk Sensor including: 1×b.safe Hook-and-Look Tape, width 10 mm × length 2 m, 1×b.safe Hook-and-Loop Strips, two-part, width 2×length 25 mm
1×Signal cable, length 2 m, connection 1: socket, connection 2: plug, 1×Signal cable, length 2 m, connections: 1 female, 2 male
Specifications
  • Description:
    Level control set, 1-channel; No. of containers to be monitored: 1