Order Entry
Germany
Orders LinkContactUsLinkComponent
 

TAPI-0 ≥95% Matrix metalloprotease (MMP) and TACE inhibitor

Catalog # ABCAAB141497-1
Supplier:   Abcam
TAPI-0 ≥95% Matrix metalloprotease (MMP) and TACE inhibitor
Catalog # ABCAAB141497-1
Supplier:  Abcam
CAS Number:  143457-40-3
TAPI-0 ≥95% Matrix metalloprotease (MMP) and TACE inhibitor
CAS Number:  143457-40-3
Price
359,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Inhibits TNF-α processing. Also a general matrix metalloprotease and TACE inhibitor.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • CAS Number:143457-40-3
  • Molecular Weight:456,5 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% TAPI-0