Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Anthrone carbohydrate determination reagent

Catalog # ABCAAB146385-25
Marketsource
Supplier:   Abcam
Anthrone carbohydrate determination reagent
Catalog # ABCAAB146385-25
Supplier:  Abcam
Marketsource
CAS Number:  90-44-8
Anthrone carbohydrate determination reagent
CAS Number:  90-44-8
Price
76,90 €
Log in for availability
1 * 25 g
Product Details & Documents

Tricyclic aromatic hydrocarbon used in the colorometric determination of carbohydrates.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Anthracen-9(10H)-one
  • CAS Number:90-44-8
  • Molecular Weight:194,23 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Anthrone