Order Entry
Germany
Orders LinkContactUsLinkComponent
Phenol red pH-indicator

Phenol red pH-indicator

Catalog # ABCAAB146336-25
Supplier:   Abcam
Phenol red pH-indicator
Catalog # ABCAAB146336-25
Supplier:  Abcam
CAS Number:  143-74-8
Phenol red pH-indicator
CAS Number:  143-74-8
Price
76,90 €
Log in for availability
1 * 25 g
Product Details & Documents

pH indicator widely used in culture media to identify pH changes from neutral (red) to acidic (yellow) values. Typically a change in colour from red to yellow indicates high rates of cellular death when used in tissue cultures. Observed to act as a weak oestrogen in the culture of breast cancer cells. Induced oogenesis from cells scraped from the surface of adult ovaries.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Molecular Weight:354,37 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Phenol red
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.