Order Entry
Germany
Orders LinkContactUsLinkComponent
Ninhydrin Amine detection reagent

Ninhydrin Amine detection reagent

Catalog # ABCAAB146213-25
Supplier:   Abcam
Ninhydrin Amine detection reagent
Catalog # ABCAAB146213-25
Supplier:  Abcam
CAS Number:  485-47-2
Ninhydrin Amine detection reagent
CAS Number:  485-47-2
Price
76,90 €
Log in for availability
1 * 25 g
Product Details & Documents

Ninhydrin reacts with a mMonia and primary and secondary amines producing a blue or purple reaction product (diketohydrindylidene-diketohydrindamine). Co mMonly used for the development of fingermarks on porous surfaces. Also useful in the analysis of free amino groups in proteins, peptides and amino acids. May be useful for detecting ninhydrin-reactive nitrogens in soil for forensic applications.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
  • CAS Number:485-47-2
  • Molecular Weight:178,14 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Ninhydrin