Order Entry
Germany
Orders LinkContactUsLinkComponent
Nuclear fast red nucleic acid stain nucleic acid stain

Nuclear fast red nucleic acid stain nucleic acid stain

Catalog # ABCAAB146372-5
Marketsource
Supplier:   Abcam
Nuclear fast red nucleic acid stain nucleic acid stain
Catalog # ABCAAB146372-5
Supplier:  Abcam
Marketsource
CAS Number:  6409-77-4
Nuclear fast red nucleic acid stain nucleic acid stain
CAS Number:  6409-77-4
Price
139,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Fast-acting biological stain that targets nucleic acids. Works more quickly than Hematoxylin. Also suitable for the determination of calcium.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL
  • CAS Number:6409-77-4
  • Molecular Weight:357,28 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Nuclear Fast Red
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.