BAPTA-AM ≥97% Ca²⁺ chelator
Product Details & Documents
Selective Ca²⁺ chelator. Cell-permeable analog of BAPTA. Useful for manipulation of intracellular free Ca²⁺ levels. Shows varied biological activity. Blocks hKv1.5, Kv11.1 (hERG) and hKv1.3 channels (Ki values are 1.23, 1.30 and 1.45 μM, respectively).
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:126150-97-8
- Molecular Weight:764,7 g/mol
Specifications
Specification Test Results
- Constituents::100% 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acidtetrakis(acetoxymethyl)ester
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



