Orders LinkContactUsLinkComponent
ToughStripe® Max Solid Floor Tapes
ToughStripe® Max Solid Floor Tapes
New Product
Supplier:   Brady
ToughStripe® Max Solid Floor Tapes
Supplier:  Brady
New Product
Certificates
ToughStripe® Max Solid Floor Tapes
ToughStripe® Max Solid Floor Tapes
30 Options Available Below
Product Details & Documents
Built stronger, lasts longer on punishing industrial floors. ToughStripe® Max floor marking tape is purpose-built to hold its ground in forklift and heavy traffic areas.
  • Strong polyester (PETG) material is four times more rigid than vinyl and contains no plasticisers
  • Matte finish provides excellent colour contrast for clear visibility under any lighting
  • Damage-resistant and impact-resistant
  • Rubber adhesive
  • Minimum application temperature: 10 °C
  • Service temperature: 10 to 60 °C
ToughStripe® Max Solid Floor Tape is re-engineered to be twice as thin yet twice as durable to better withstand damaging impacts in forklift and heavy traffic areas. The thinner profile and true edge-to-edge rubber adhesive minimise pick points. The damage-isolating design keeps tape intact longer by preventing rips, gouges, and tears from running.
Compact construction results in a smaller, lightweight roll, which can be easily installed by one person with minimal tools. This floor tape is compatible with the ToughStripe® Floor Tape Applicator (part number BRDY150852).
Product Family Options
Product Information
  • Colour
    Length
    Width
    Thickness
    Availability
    Price
  • BRDY177038
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Black
    30,48 m
    50,80 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
    Specifications:
    New Product
    New Product
  • BRDY177019
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Blue
    30,48 m
    50,80 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
    Specifications:
    New Product
    New Product
  • BRDY177068
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Brown
    30,48 m
    50,80 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
    Specifications:
    New Product
    New Product
  • BRDY177022
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Green
    30,48 m
    50,80 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
    Specifications:
    New Product
    New Product
  • BRDY177062
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Grey
    30,48 m
    50,80 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
    Specifications:
    New Product
    New Product
  • BRDY177030
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Orange
    30,48 m
    50,80 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
    Specifications:
    New Product
    New Product
  • BRDY177045
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Purple
    30,48 m
    50,80 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
    Specifications:
    New Product
    New Product
  • BRDY177020
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Red
    30,48 m
    50,80 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
    Specifications:
    New Product
    New Product
  • BRDY177021
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    White
    30,48 m
    50,80 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
    Specifications:
    New Product
    New Product
  • BRDY177015
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Yellow
    30,48 m
    50,80 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
    Specifications:
    New Product
    New Product
  • BRDY177049
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Black
    30,48 m
    76,20 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,71 €
    196,71 €
    1 * 1 ST
       196,71 €
    Specifications:
    New Product
    New Product
  • BRDY177023
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Blue
    30,48 m
    76,20 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,71 €
    196,71 €
    1 * 1 ST
       196,71 €
    Specifications:
    New Product
    New Product
  • BRDY177070
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Brown
    30,48 m
    76,20 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,71 €
    196,71 €
    1 * 1 ST
       196,71 €
    Specifications:
    New Product
    New Product
  • BRDY177031
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Green
    30,48 m
    76,20 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,71 €
    196,71 €
    1 * 1 ST
       196,71 €
    Specifications:
    New Product
    New Product
  • BRDY177078
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Grey
    30,48 m
    76,20 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,71 €
    196,71 €
    1 * 1 ST
       196,71 €
    Specifications:
    New Product
    New Product
  • BRDY177034
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Orange
    30,48 m
    76,20 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,71 €
    196,71 €
    1 * 1 ST
       196,71 €
    Specifications:
    New Product
    New Product
  • BRDY177043
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Purple
    30,48 m
    76,20 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,71 €
    196,71 €
    1 * 1 ST
       196,71 €
    Specifications:
    New Product
    New Product
  • BRDY177028
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Red
    30,48 m
    76,20 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,71 €
    196,71 €
    1 * 1 ST
       196,71 €
    Specifications:
    New Product
    New Product
  • BRDY177026
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    White
    30,48 m
    76,20 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,71 €
    196,71 €
    1 * 1 ST
       196,71 €
    Specifications:
    New Product
    New Product
  • BRDY177014
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Yellow
    30,48 m
    76,20 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,71 €
    196,71 €
    1 * 1 ST
       196,71 €
    Specifications:
    New Product
    New Product
  • BRDY177059
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Black
    30,48 m
    101,60 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,84 €
    223,84 €
    1 * 1 ST
       223,84 €
    Specifications:
    New Product
    New Product
  • BRDY177037
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Blue
    30,48 m
    101,60 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,84 €
    223,84 €
    1 * 1 ST
       223,84 €
    Specifications:
    New Product
    New Product
  • BRDY177069
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Brown
    30,48 m
    101,60 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,84 €
    223,84 €
    1 * 1 ST
       223,84 €
    Specifications:
    New Product
    New Product
  • BRDY177036
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Green
    30,48 m
    101,60 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,84 €
    223,84 €
    1 * 1 ST
       223,84 €
    Specifications:
    New Product
    New Product
  • BRDY177096
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Grey
    30,48 m
    101,60 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,84 €
    223,84 €
    1 * 1 ST
       223,84 €
    Specifications:
    New Product
    New Product
  • BRDY177042
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Orange
    30,48 m
    101,60 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,84 €
    223,84 €
    1 * 1 ST
       223,84 €
    Specifications:
    New Product
    New Product
  • BRDY177074
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Purple
    30,48 m
    101,60 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,84 €
    223,84 €
    1 * 1 ST
       223,84 €
    Specifications:
    New Product
    New Product
  • BRDY177032
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Red
    30,48 m
    101,60 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,84 €
    223,84 €
    1 * 1 ST
       223,84 €
    Specifications:
    New Product
    New Product
  • BRDY177027
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    White
    30,48 m
    101,60 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,84 €
    223,84 €
    1 * 1 ST
       223,84 €
    Specifications:
    New Product
    New Product
  • BRDY177016
    ToughStripe® Max solid floor tape, polyester
    ToughStripe® Max solid floor tape, polyester
    Yellow
    30,48 m
    101,60 mm
    0,559 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,84 €
    223,84 €
    1 * 1 ST
       223,84 €
    Specifications:
    New Product