10X RIPA Buffer, Abcam
Product Details & Documents
10X RIPA lysis buffer (pH 7.5) is a means of cell lysis and protein solubilization for both adherent and suspension-cultured mammalian cells. - Cited in over 180 publications - Suitable for immunoprecipitation (IP), ELISA, SDS-PAGE, western blot - Effectively extracts cytoplasmic, nuclear and membrane proteins
- pH: 7.5 Constituents: 10% Nonylphenol, ethoxylated, 8.8% Sodium chloride, 6.1% Tris, 5% Sodium deoxycholate, 1.12% Sodium pyrophosphate decahydrate, 1% Sodium lauryl sulfate, 0.38% EGTA, 0.29% EDTA, 0.22% disodium;1,3-dihydroxypropan-2-yl phosphate;hydrate, 0.18% Sodium orthovanadate
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
Specifications
- Description:10X RIPA Buffer
- Cat. No.:ABCAAB156034-15
- Storage temperature:Store at Ambient.



