Chromatin Extraction Kit, Abcam
Product Details & Documents
60-minute chromatin extraction kit is a complete set of optimized buffers and reagents for isolating chromatin or DNA-protein complex from mammalian cells and tissues in a simple and rapid format. - Cited in over 50 publications
Achtung! Bitte beachten: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Spezifikationen:
Spezifikationen:
- Description:Chromatin Extraction Kit
- Cat. No.:ABCAAB117152-100
- Storage temperature:Store at Multi.
Spezifikationen:
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.