Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Sodium azide solution 1%

Sodium azide solution 1%
Supplier:  G-Biosciences
Certificates
Sodium azide solution 1%

3 Options Available Below

Product Details & Documents

G-Biocide-III is sodium azide based preservative. Azide anions bind to trivalent iron in porphyrin complexes and inhibit the activity of catalase, peroxidases and cytochrome oxidases. It acts as bacteriostatic by inhibiting cytochrome oxidase.

  • CAS number: 26628-22-8 Molecular formula: NaN3 Molecular weight: 65.0099 InChI Key: PXIPVTKHYLBLMZ-UHFFFAOYSA-N Recommended concentration for use: 0.02 to 0.05%

A 1% aqueous solution of sodium azide [154 mm] for use as a preservative for laboratory reagents. The use of the 1% solution is convenient and minimizes exposure and contamination risks normally encountered while handling sodium azide powder. Used as preservative to preserve wide range of research reagents such as buffers, blocking solutions for ELISA, chromatography resins and Immunoglobulins storage solutions.

Synonyms:  Sodium triazide
  • CAS Number:26628-22-8
  • Formula:NaN₃
  • MDL Number:MFCD00003536
  • Molecular Weight:65.01 g/mol

 

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 36,70 €
    36,70 €
    1 * 100 mL
       36,70 €

  • Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 80,10 €
    80,10 €
    1 * 250 mL
       80,10 €

  • Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 146,00 €
    146,00 €
    1 * 500 mL
       146,00 €