Order Entry
Germany
Orders LinkContactUsLinkComponent
Phenol Red solution 0,5%

Phenol Red solution 0,5%

Phenol Red solution 0,5%
Supplier:  G-Biosciences
Certificates
Phenol Red solution 0,5%

2 Options Available Below

Product Details & Documents

Phenol Red Solution [0.5%] is a pH indicator commonly used in cell culture and biochemical applications.

  • Synonym: Phenolsulfonphthalein CAS number: 143-74-8 Molecular formula: C19H14O5S Molecular weight: 354.38

At 0.5% concentration, it provides a visible color change across a physiological pH range, allowing quick assessment of pH shifts in media, buffers, or experimental reactions.

Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354.38 g/mol

Documents

Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.