ContactUsLinkComponent
Washing Bottle Lower Part for 4010*, 4209* or 4213*
Washing Bottle Lower Part for 4010*, 4209* or 4213*
Catalog # ROBU41110
Supplier:  ROBU GLASFILTER
undefined
Washing Bottle Lower Part for 4010*, 4209* or 4213*
Catalog # ROBU41110
Supplier:  ROBU GLASFILTER
Specifications
  • Description:
    Washing bottle lower part, GL 45
  • For bottles:
    Fits 4110*, 4209* and 4213*
Specifications
About this item
ROBU® washing bottle lower part, made from durable borosilicate glass, serves as the liquid reservoir in gas absorption processes.
  • Borosilicate glass
  • High chemical and thermal resistance
  • High-quality GL 45 thread
  • Robust and reusable
  • Ideal for use with VitraPOR® heads
  • Clear visibility of contents
The lower part of the ROBU Washing Bottle is crafted from high-quality borosilicate glass, ensuring exceptional resistance to chemicals and thermal stress. Designed to serve as the receiving vessel in gas absorption systems, it securely holds liquids while allowing visual monitoring of the absorption process. Compatible with ROBU heads with standard joints and VitraPOR filter discs, this lower part forms a reliable, durable solution for efficient gas scrubbing applications in laboratories and industrial settings.