Orders LinkContactUsLinkComponent
Micro-Filter Candles
Supplier:   ROBU GLASFILTER
Micro-Filter Candles
Supplier:  ROBU GLASFILTER
Certificates
Micro-Filter Candles
Micro-Filter Candles
28 Options Available Below
Product Details & Documents
VitraPOR® micro-filter candles provide precise filtration in compact setups. Ideal for low-volume applications requiring chemical resistance and fine particle retention.
  • VitraPOR® porous glass filter
  • Fine and uniform pore structure
  • Compact size
  • High chemical and thermal resistance
  • Suitable for gases and liquids
  • Reusable
  • Ideal for laboratory and process applications
ROBU Micro-Filter Candles with integrated VitraPOR filter are high-precision filtration elements designed for small-volume and confined-space applications. Made entirely of borosilicate glass with an integrated VitraPOR structure, they deliver consistent and efficient particle separation for both liquids and gases. Their high chemical resistance and thermal stability make them ideal for use in analytical laboratories, chemical processing, and other demanding environments. These micro-filter candles are durable, autoclavable, and can be easily regenerated through cleaning or thermal treatment.
 : In compliance with ISO 4796-80 and ASTM E128-99 standards.
Product Family Options
Product Information
  • Porosity
    Ø×H
    Tube ext.Ø
    Tube length
    Availability
    Price
  • ROBU18140
    Micro-filter candle
    Micro-filter candle
    0
    20×55 mm
    20 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 370,00 €
    370,00 €
    1 * 5 ST
       370,00 €
  • ROBU18120
    Micro-filter candle
    Micro-filter candle
    0
    13×25 mm
    8 mm
    210 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 183,00 €
    183,00 €
    1 * 5 ST
       183,00 €
  • ROBU18110
    Micro-filter candle
    Micro-filter candle
    0
    9×20 mm
    8 mm
    210 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 183,00 €
    183,00 €
    1 * 5 ST
       183,00 €
  • ROBU18100
    Micro-filter candle
    Micro-filter candle
    0
    6×15 mm
    6 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 183,00 €
    183,00 €
    1 * 5 ST
       183,00 €
  • ROBU18130
    Micro-filter candle
    Micro-filter candle
    0
    20×30 mm
    20 mm
    180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 310,00 €
    310,00 €
    1 * 5 ST
       310,00 €
  • ROBU18131
    Micro-filter candle
    Micro-filter candle
    1
    20×30 mm
    20 mm
    180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 310,00 €
    310,00 €
    1 * 5 ST
       310,00 €
  • ROBU18141
    Micro-filter candle
    Micro-filter candle
    1
    20×55 mm
    20 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 370,00 €
    370,00 €
    1 * 5 ST
       370,00 €
  • ROBU18101
    Micro-filter candle
    Micro-filter candle
    1
    6×15 mm
    6 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 183,00 €
    183,00 €
    1 * 5 ST
       183,00 €
  • ROBU18111
    Micro-filter candle
    Micro-filter candle
    1
    9×20 mm
    8 mm
    210 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 183,00 €
    183,00 €
    1 * 5 ST
       183,00 €
  • ROBU18121
    Micro-filter candle
    Micro-filter candle
    1
    13×25 mm
    8 mm
    210 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 183,00 €
    183,00 €
    1 * 5 ST
       183,00 €
  • ROBU18142
    Micro-filter candle
    Micro-filter candle
    2
    20×55 mm
    20 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 370,00 €
    370,00 €
    1 * 5 ST
       370,00 €
  • ROBU18112
    Micro-filter candle
    Micro-filter candle
    2
    9×20 mm
    8 mm
    210 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 183,00 €
    183,00 €
    1 * 5 ST
       183,00 €
  • ROBU18102
    Micro-filter candle
    Micro-filter candle
    2
    6×15 mm
    6 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 183,00 €
    183,00 €
    1 * 5 ST
       183,00 €
  • ROBU18122
    Micro-filter candle
    Micro-filter candle
    2
    13×25 mm
    8 mm
    210 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 183,00 €
    183,00 €
    1 * 5 ST
       183,00 €
  • ROBU18132
    Micro-filter candle
    Micro-filter candle
    2
    20×30 mm
    20 mm
    180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 310,00 €
    310,00 €
    1 * 5 ST
       310,00 €
  • ROBU18143
    Micro-filter candle
    Micro-filter candle
    3
    20×55 mm
    20 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 370,00 €
    370,00 €
    1 * 5 ST
       370,00 €
  • ROBU18133
    Micro-filter candle
    Micro-filter candle
    3
    20×30 mm
    20 mm
    180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 310,00 €
    310,00 €
    1 * 5 ST
       310,00 €
  • ROBU18103
    Micro-filter candle
    Micro-filter candle
    3
    6×15 mm
    6 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 183,00 €
    183,00 €
    1 * 5 ST
       183,00 €
  • ROBU18113
    Micro-filter candle
    Micro-filter candle
    3
    9×20 mm
    8 mm
    210 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 183,00 €
    183,00 €
    1 * 5 ST
       183,00 €
  • ROBU18123
    Micro-filter candle
    Micro-filter candle
    3
    13×25 mm
    8 mm
    210 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 183,00 €
    183,00 €
    1 * 5 ST
       183,00 €
  • ROBU18144
    Micro-filter candle
    Micro-filter candle
    4
    20×55 mm
    20 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 421,00 €
    421,00 €
    1 * 5 ST
       421,00 €
  • ROBU18134
    Micro-filter candle
    Micro-filter candle
    4
    20×30 mm
    20 mm
    180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 346,00 €
    346,00 €
    1 * 5 ST
       346,00 €
  • ROBU18114
    Micro-filter candle
    Micro-filter candle
    4
    9×20 mm
    8 mm
    210 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 201,00 €
    201,00 €
    1 * 5 ST
       201,00 €
  • ROBU18104
    Micro-filter candle
    Micro-filter candle
    4
    6×15 mm
    6 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 200,00 €
    200,00 €
    1 * 5 ST
       200,00 €
  • ROBU18124
    Micro-filter candle
    Micro-filter candle
    4
    13×25 mm
    8 mm
    210 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 202,00 €
    202,00 €
    1 * 5 ST
       202,00 €
  • ROBU18115
    Micro-filter candle
    Micro-filter candle
    5
    9×20 mm
    8 mm
    210 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 389,00 €
    389,00 €
    1 * 5 ST
       389,00 €
  • ROBU18105
    Micro-filter candle
    Micro-filter candle
    5
    6×15 mm
    6 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 317,00 €
    317,00 €
    1 * 5 ST
       317,00 €
  • ROBU18125
    Micro-filter candle
    Micro-filter candle
    5
    13×25 mm
    8 mm
    210 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 471,00 €
    471,00 €
    1 * 5 ST
       471,00 €