Order Entry
Germany
ContactUsLinkComponent
PFAS 24 Calibration Standard
PFAS 24 Calibration Standard
Catalog # REST30733
Supplier:  Restek
New Product
CAS Number:  
PFAS 24 Calibration Standard
Catalog # REST30733
Supplier:  Restek
New Product
CAS Number:  

Specifications

  • Concentration:
    1 µg/ml, methanol (1 mM KOH)
  • Description:
    PFAS 24 calibration standard
  • Shipping conditions:
    Ambient
  • Shelf life:
    6 - 60 months (on ship date)
  • Storage temperature:
    0 °C or colder
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The 24-component mix simplifies calibration complexity, saving time and reducing cost.

  • Verified composition and stability ensure long-lasting shelf life

    Two independently produced lots available

    Ideal for the analysis of PFAS in water and other matrices

Components:

1H,1H,2H,2H-Perfluorodecane sulfonic acid (8:2 FTS) (39108-34-4)
1H,1H,2H,2H-Perfluorohexane sulfonic acid (4:2 FTS) (757124-72-4)
1H,1H,2H,2H-Perfluorooctane sulfonic acid (6:2 FTS) (27619-97-2)
N-ethylperfluoro-1-octanesulfonamidoacetic acid (NEtFOSAA) (2991-50-6)
N-methylperfluoro-1-octanesulfonamidoacetic acid (NMeFOSAA) (2355-31-9)
Perfluoro-1-decanesulfonic acid (PFDS) (335-77-3)
Perfluoro-1-nonanesulfonic acid (PFNS) (68259-12-1)
Perfluoro-1-octanesulfonamide (FOSA) (754-91-6)
Perfluoro-1-pentanesulfonic acid (PFPeS) (2706-91-4)
Perfluorobutanesulfonic acid (PFBS) (375-73-5)
Perfluorobutanoic acid (PFBA) (375-22-4)
Perfluorodecanoic acid (PFDA) (335-76-2)
Perfluorododecanoic acid (PFDOA) (307-55-1)
Perfluoroheptanesulfonic acid (PFHpS) (375-92-8)
Perfluoroheptanoic acid (PFHpA) (375-85-9)
Perfluorohexanesulfonic acid (PFHxS)* ( 355-46-4)
Perfluorohexanoic acid (PFHxA) (307-24-4)
Perfluorononanoic acid (PFNA) (375-95-1)
Heptadecafluorooctanesulfonic acid (PFOS)* (1763-23-1)
Perfluorooctanoic acid (PFOA)* (335-67-1)
Perfluoropentanoic acid (PFPeA) (2706-90-3)
Perfluorotetradecanoic acid (PFTeDA) (376-06-7)
Perfluorotridecanoic acid (PFTrDA) (72629-94-8)
Perfluoroundecanoic acid (PFUnA) (2058-94-8)

Certifications:

Certified reference material (CRM). Meets EPA 8327 method requirements.