- Sterile:+
- Colour:Natural
- Description:Cleanroom 96-well Polypropylene Sample Processing & Storage Plates
- Pk:Bulk
- Well volume:330 μL
Thermo Scientific Abgene 96 Well Polypropylene Storage Microplates provide excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. All plates are ANSI format for compatibility with automated systems and are manufactured in our cleanroom facility to ensure molecular grade quality. To further assist in storage and tracking, all plates can be supplied with custom barcoding.
Alphanumeric grid-referencing and industry standard footprint
Sealing options: Adhesive Seals, Heat Seals and Cap Strips
Cleanroom manufactured
Aplication: Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation.
For use with equipment: ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers.
Fit for Purpose: PCR Ready
Format: Standard
Array: 8×12
Plate: SizeANSI/ SLAS Standard
Max.RCF: 5000×g
Autoclavable: Autoclavable
Binding Type: Low binding PP
Certified RNase, DNase, PCR Inhibitors, and human DNA Free.
ANSI Compliant.
One per pack
100 per case