Flat Gasket Seals
Catalog # LEYBES23955165
Supplier: LEYBOLD
New Product
Specifications
- Description:Flat seal oil inlet/oil drain
Specifications
About this item
These oil drain gaskets are designed to seal the oil drain plug on Leybold D40B and D65B rotary vane vacuum pumps.