Immobilised Catalase
Catalog # GENO786-1906
Supplier: G-Biosciences
New Product
CAS Number:
Specifications
- Description:Immobilized catalase, 2 ml resin
Specifications
About this item
Immobilised Catalase is prepared by covalent coupling of catalase enzyme to 6% cross-linked agarose beads. Immobilised Catalase is used to remove hydrogen peroxide from biological samples for use in downstream applications which require absenceof hydrogen peroxide as it causes interference.
Catalase coupled to 6% cross-linked Agarose
Extent of labeling is 1 to 2 mg calatalse per ml resin
Shipped as hydrated resin in a 50% Glycerol-PBS buffer
Available in 2 ml size
Immobilised Catalase can be used to remove hydrogen peroxide in wide range of samples including urine, serum, plasma and cellular extracts.