Click-On In-Line Super Clean® Replacement Gas Traps
Supplier: Restek
New Product
Certificates
About this item
Super-clean click-on traps measure to be 21,6×3,2 cm (8,5×1,25").
Triple trap - removes oxygen, moisture, and hydrocarbons
Super-clean click-on traps measure to be 21,6×3,2 cm (8,5×1,25").
Triple trap - removes oxygen, moisture, and hydrocarbons