ContactUsLinkComponent
Tork W8 Heavy-Duty Cleaning Cloths
Tork W8 Heavy-Duty Cleaning Cloths
Catalog # SCAA530150
Supplier:  Essity
New Product
undefined
Tork W8 Heavy-Duty Cleaning Cloths
Catalog # SCAA530150
Supplier:  Essity
New Product
Specifications
  • Packed:
    8×45 cloths
  • Colour:
    White
  • Description:
    Cloths, refill, premium, compatible with Tork W8 dispensers
  • L×W:
    193×160 mm (Folded size), 385×320 mm (Un folded size)
Specifications
About this item
Protect hands from heat and metal scraps with heavy-duty design. Tackle wiping tasks that require strength in no time with this multipurpose cleaning cloth.
  • Reduce waste - durable and reusable cleaning cloth won't tear easily Swiftly remove oil, grease, lubricants and dirt, thanks to its high absorbency
Workspaces can become crowded with paper rolls and rental cloths - multipurpose cleaning cloth can handle a wide range of tasks, saving the time and money. This tough design is suitable for use with most solvents for further versatility. Removing oil, lubricants and dirt is easy, thanks to its high absorbency. Plus, the heavy-duty material shields hands from heat and metal scraps for safer working. Finished with an exelCLEAN® effect, these cleaning cloths work to achieve a professional-looking result.
Certifications: FSC MIX Credit, SA-COC-008266.