ContactUsLinkComponent
Tork Light Duty Cleaning Cloths
Tork Light Duty Cleaning Cloths
Catalog # SCAA500479
Supplier:  Essity
New Product
undefined
Tork Light Duty Cleaning Cloths
Catalog # SCAA500479
Supplier:  Essity
New Product
Specifications
  • Packed:
    4 packs, 120 cloths each (4×120 cloths)
  • Colour:
    White
  • Description:
    Cloths, refill, premium, compatible with Tork W4 dispensers
  • L×W:
    108×355 mm (Folded size), 415×355 mm (Un folded size)
Specifications
About this item
Multipurpose cleaning cloths for light-duty cleaning tasks. Use these durable cloths wet or dry to clean dust and dirt, and can rinse and reuse them as needed.
  • These disposable cleaning cloths are compatible with most chemical solvents, so no need to change the cleaning products
    Compatible Top-pak dispenser has one-at-a-time dispensing to help optimise consumption and minimise waste
Tork light-duty cleaning cloths are an economical, multipurpose wiper. Compatible with most chemical solvents, these durable cloths can be used wet or dry for various light-duty cleaning tasks. They’re soft and flexible, making it easy to clean in hard-to-reach places, and more durable than a paper wiper, so can rinse and reuse them. The compatible Top-pak dispenser delivers one-at-a-time dispensing to minimise waste.
Certifications: FSC MIX Credit, SA-COC-008266.