ContactUsLinkComponent
Tork W8 Industrial Cleaning Cloths
Tork W8 Industrial Cleaning Cloths
Catalog # SCAA520350
Supplier:  Essity
New Product
undefined
Tork W8 Industrial Cleaning Cloths
Catalog # SCAA520350
Supplier:  Essity
New Product
Specifications
  • Packed:
    8 packs, 55 cloths (8×55 cloths)
  • Colour:
    Grey
  • Description:
    Cloths, refill, folded, compatible with Tork W8 dispensers
  • L×W:
    158,75×193,802 mm (Folded size), 384,9878×319,9892 mm (Un folded size)
Specifications
About this item
Easily wipe away oil, grease, lubricants and dirt with its strong texture and high absorbency. Avoid wasting time by using only one cloth for light-to-medium industrial tasks.
  • Get rid of dirt in tight spaces, thanks to the soft and flexible feel
    Save up to 41% of solvents with the exelCLEAN® material, looking after health and the environment
Get rid of hard-to-shift oil and grease with the strong and highly absorbent cloths. Dirt in tight spaces can be hard to tackle, but Tork industrial cleaning cloths are soft and flexible to make easy work of small corners. This multipurpose design produces up to 85% less waste than traditional rags for cost-effective use. Plus, the professional exelCLEAN® material saves up to 41% of solvents, helping health and the environment. Use these cloths for all light-to-medium industrial tasks.
Certifications: Health Canada