Specifications
- Packed:4 buckets, 58 sheets (4×58 wipes)
- Colour:White
- Description:Cleaning wet wipes, refill, premium, compatible with Tork W14 dispensers
- L×W:15660×135 mm
Specifications
About this item
Clean hands effectively with these professional-quality wipes.
Remove oil, grease, lubricants and dirt with ease, thanks to the strong texture and high absorbency of these wet wipes
Avoid potential irritations with wipes that are free of fragrance and kind to the skin
Refilling is easy and efficient with the bucket-style dispenser
When there is no access to water, these Tork hand cleaning wet wipes can be used. Their strong texture works to swiftly remove oil, grease and lubricants from hands. It can tackle dirt effectively, too, thanks to their professional cleaning ability. Tough on dirt doesn't mean tough on skin as the premium 1-ply wipes have a gentle, fragrance-free formula. Plus, refilling is less of a chore - the wipes together with this bucket makes the process hassle-free and efficient.