Orders LinkContactUsLinkComponent
Gas Sampling Balloons
Gas Sampling Balloons
Supplier:   LAMAPLAST
Gas Sampling Balloons
Supplier:  LAMAPLAST
Certificates
Gas Sampling Balloons
Gas Sampling Balloons
5 Options Available Below
Product Details & Documents
Gas sampling balloons are made from natural latex.
  • Volume ranges from 1500 to 8000 ml
  • Colour: pale yellow/red
The 1500 ml balloons have a blowpipe diameter of 4,9 mm, with a blowpipe length of 95 mm. The 2000 and 4000 ml balloons have a blowpipe diameter of 5,85 mm, with a blowpipe length of 86 mm. The 5000 and 8000 ml balloons have a blowpipe diameter of 4,8 mm, with a blowpipe length of 93 mm.
Packaging: Each ballon is packaged in a single plastic bag.
Documents
Product Family Options
Product Information
  • Ø
    Volume
    Availability
    Price
  • LAMA80601
    Gas sampling balloon
    Gas sampling balloon
    150 mm
    1500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 60,20 €
    60,20 €
    1 * 1 ST
       60,20 €
  • LAMA80602
    Gas sampling balloon
    Gas sampling balloon
    160 mm
    2000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 71,80 €
    71,80 €
    1 * 1 ST
       71,80 €
  • LAMA80603
    Gas sampling balloon
    Gas sampling balloon
    200 mm
    4000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,70 €
    82,70 €
    1 * 1 ST
       82,70 €
  • LAMA80604
    Gas sampling balloon
    Gas sampling balloon
    230 mm
    5000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
  • LAMA80605
    Gas sampling balloon
    Gas sampling balloon
    250 mm
    8000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 134,00 €
    134,00 €
    1 * 1 ST
       134,00 €