Specifications
- Description:TopStream 400 Safe In extraction system, cell culture medium suction system
Specifications
About this item
The TopStream 400 Safe In is used for extraction specifically for the BSC Herasafe from Thermo Scientific.
The suction pipettes are autoclavable and suitable for all standard tips 200 and 1000 µl, and Pasteur pipettes and aspiration pipettes can also be attached
- The liquid pump can also be used to transport liquid from A to B
- The pump is suitable for sterile filtration with bottle-top filters
- Pump made of high-quality aluminum housing, encapsulated, suitable for surface disinfection and with UV protection
- Excellent personal protection and environmental protection
- Small, mobile, space-saving pump module
The TopStream 400 Safe In liquid pump is used for delicate and large volume suction of cell culture medium and laboratory liquids. The system has been specially developed for the Herasafe biological safety workbench from Thermo Scientific. The pump technology allows the connection of standard laboratory glass bottles that are already available in every biological laboratory. Furthermore, 5 L Bürkle flat tanks can be combined with the pump and workbench. Any desired plastic containers can also be connected as a collecting container. The pump system, including the pump head, can be disinfected with a Bacillol AF or 80% ethanol disinfectant solution. The system is thus chemically sterilised and prevents contamination and infections. With the perfect aerosol protection, the system maintains the safe air flow of the safety workbench and thus personal protection. EN 12469 for safety workbenches is thus complied with. Various pump module holders for the safety workbench are available as accessories. The pump can be plugged directly into the safety cabinet's socket using a special holder.
The suction pipettes are autoclavable and are suitable for all standard tips 200 µl and 1000 µl, and Pasteur pipettes and aspiration pipettes can also be plugged in. The liquid pump can also be used to transport liquid from A to B. The pump is suitable for sterile filtration with bottle-top filters.
W×D×H: 114×50×105 mm
Weight: 0,5 kg
Various pump module holders for the safety workbench are available as accessories. Contact your Avantor representative for more information.
Includes one liquid pump with hose set, adapter/particle filter for Pasteur, tips 1000 µl and serological pipettes.