ContactUsLinkComponent
VALEGRO Recirculating Coolers
VALEGRO Recirculating Coolers
Supplier:  JULABO GmbH
New Product
Certificates
undefined
VALEGRO Recirculating Coolers
Supplier:  JULABO GmbH
New Product
Specifications
  • Catalog No:
  • JULA9610035
  • 473-3201
  • Model:
  • VALEGRO 350
  • VALEGRO 500
  • Pump flow rate:
  • 16 L/min
  • 19 L/min
  • Dimensions:
  • 23×45×45 cm
  • 23×45×45 cm
Specifications
Option
Product Information