Steridon Sterile Coveralls with Hood
Supplier: ISOFIELD IRELAND
New Product
Certificates
About this item
The tri-laminate Steridon cleanroom coverall is aseptically folded and can be donned in 60 seconds or less.
Sterile cleanroom coverall with hood
- Latex-free, enclosed and elasticated face, wrists, and ankles
- Ergo-fit design at back and shoulders allowing greater range of motion
- Aseptically presented fold
- Colour-coded left and right inner tabs to facilitate donning
- Removable external tabs at zipper to ensure sterile field is maintained
- Validated for sterility by X-Ray
- Sterility Assurance Level (SAL) 10⁻⁶
Steridon cleanroom coveralls are validated sterile via X-ray and tested against chemo to ASTM F 739. The anti-static, low-linting, viral barrier provides essential protection in Grade A/B environments for all cleanroom activities.
Packaging:
Coveralls are packaged 1 piece per sealed inner bag, 1 inner bag per sealed outer bag, 20 outer bags per carton liner with twist tie, 20 pieces per carton (XXS to XXL); 15 pieces per carton (3XL to 5XL).
Specifications
- Catalog No:
- ISOF80111M
- ISOF80111S
- ISOF80111XS
- ISOF80111XXS
- ISOF80111L
- ISOF80111XL
- ISOF80111XXL
- ISOF80111XXXL
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Cleanroom class:
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Particle shedding (Helmke Drum):
- 160 counts/min for particles larger than 0,3 µm
- 160 counts/min for particles larger than 0,3 µm
- 160 counts/min for particles larger than 0,3 µm
- 160 counts/min for particles larger than 0,3 µm
- 160 counts/min for particles larger than 0,3 µm
- 160 counts/min for particles larger than 0,3 µm
- 160 counts/min for particles larger than 0,3 µm
- 160 counts/min for particles larger than 0,3 µm