VWR® Air Valve
590-0025
- Description:VWR® air valve, UNF 1/4" 28G connection
Valve housing with connection made of black PP (thread UNF 1/4" 28G) suitable for VWR safety caps.
6 month service life
With integrated check valve and upstream membrane made of porous PTFE (1 μm), including labelling field.