SUPL104726
1047261000.
: Merck
New Product
:
- Pack type:Borosilicate glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- EPA 533/537.1 Tested:Conforms
- UHPLC Gradient (at 210 nm):≤2 mAU
- Evaporation Residue:≤0.7 mg/l
- UHPLC Gradient (at 254 nm):≤0.5 mAU
- (PEG) Signal-to-Noise Ratio:≤50
:
:41.05 g/mol
:1648
:01-2119471307-38-XXXX
:GHS02
:GHS07
:Room Temperature
:75-05-8
:C₂H₃N
:MFCD00001878