Human Recombinant FGF2 (from E. coli)
About this item
Recombinant human FGF2 protein is a Human Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, FuncS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
4 Options Available Below
Product Details & Documents
Recombinant human FGF2 protein is a Human Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, FuncS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- ABCAAB61845-10
- ABCAAB61845-50
- ABCAAB61845-100
- ABCAAB61845-1
- Protein/peptide name:
- FGF2
- FGF2
- FGF2
- FGF2
- Protein synonyms:
- FGFB|Fibroblast growth factor 2|FGF-2|Basic fibroblast growth factor|Heparin-binding growth factor 2|bFGF|HBGF-2
- FGFB|Fibroblast growth factor 2|FGF-2|Basic fibroblast growth factor|Heparin-binding growth factor 2|bFGF|HBGF-2
- FGFB|Fibroblast growth factor 2|FGF-2|Basic fibroblast growth factor|Heparin-binding growth factor 2|bFGF|HBGF-2
- FGFB|Fibroblast growth factor 2|FGF-2|Basic fibroblast growth factor|Heparin-binding growth factor 2|bFGF|HBGF-2
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Animal-free:
- N
- N
- N
- N
- Biological activity:
- Standard
- Standard
- Standard
- Standard
- Purity:
- >95%
- >95%
- >95%
- >95%
- Form:
- Lyophilised
- Lyophilised
- Lyophilised
- Lyophilised
- Formulation:
- Constituents: 0,164% Sodium phosphate
- Constituents: 0,164% Sodium phosphate
- Constituents: 0,164% Sodium phosphate
- Constituents: 0,164% Sodium phosphate
- Storage conditions:
- Store at −20 °C
- Store at −20 °C
- Store at −20 °C
- Store at −20 °C
- Shipping temperature:
- 20
- 20
- 20
- 20
- Tested applications:
- FuncS, SDS-PAGE
- FuncS, SDS-PAGE
- FuncS, SDS-PAGE
- FuncS, SDS-PAGE
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- Specifications:Marketsource
More
Specifications
Cat. No.ABCAAB61845-10Protein/peptide nameFGF2Protein synonymsFGFB|Fibroblast growth factor 2|FGF-2|Basic fibroblast growth factor|Heparin-binding growth factor 2|bFGF|HBGF-2Protein/peptide typeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedAnimal-freeNBiological activityStandardPurity>95%FormLyophilisedFormulationConstituents: 0,164% Sodium phosphateStorage conditionsStore at −20 °CShipping temperature20Tested applicationsFuncS, SDS-PAGEMarketsource - Specifications:Marketsource
More
Specifications
Cat. No.ABCAAB61845-50Protein/peptide nameFGF2Protein synonymsFGFB|Fibroblast growth factor 2|FGF-2|Basic fibroblast growth factor|Heparin-binding growth factor 2|bFGF|HBGF-2Protein/peptide typeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedAnimal-freeNBiological activityStandardPurity>95%FormLyophilisedFormulationConstituents: 0,164% Sodium phosphateStorage conditionsStore at −20 °CShipping temperature20Tested applicationsFuncS, SDS-PAGEMarketsource - Specifications:Marketsource
More
Specifications
Cat. No.ABCAAB61845-100Protein/peptide nameFGF2Protein synonymsFGFB|Fibroblast growth factor 2|FGF-2|Basic fibroblast growth factor|Heparin-binding growth factor 2|bFGF|HBGF-2Protein/peptide typeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedAnimal-freeNBiological activityStandardPurity>95%FormLyophilisedFormulationConstituents: 0,164% Sodium phosphateStorage conditionsStore at −20 °CShipping temperature20Tested applicationsFuncS, SDS-PAGEMarketsource - Specifications:Marketsource
More
Specifications
Cat. No.ABCAAB61845-1Protein/peptide nameFGF2Protein synonymsFGFB|Fibroblast growth factor 2|FGF-2|Basic fibroblast growth factor|Heparin-binding growth factor 2|bFGF|HBGF-2Protein/peptide typeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedAnimal-freeNBiological activityStandardPurity>95%FormLyophilisedFormulationConstituents: 0,164% Sodium phosphateStorage conditionsStore at −20 °CShipping temperature20Tested applicationsFuncS, SDS-PAGEMarketsource
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



