Human Recombinant AP1S2 (from E. coli)
Recombinant Human AP1S2 protein is a Human Full Length protein, in the 1 to 157 aa range, expressed in Escherichia coli, with >85% purity and suitable for SDS-PAGE, MS.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Recombinant Human AP1S2 protein is a Human Full Length protein, in the 1 to 157 aa range, expressed in Escherichia coli, with >85% purity and suitable for SDS-PAGE, MS.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- Catalog No:
- ABCAAB108122-50
- ABCAAB108122-250
- Protein/peptide name:
- AP1S2
- AP1S2
- Protein synonyms:
- DC22|AP-1 complex subunit sigma-2|Adaptor protein complex AP-1 subunit sigma-1B|Adaptor-related protein complex 1 subunit sigma-1B|Clathrin assembly protein complex 1 sigma-1B small chain|Golgi adaptor HA1/AP1 adaptin sigma-1B subunit|Sigma 1B subunit of AP-1 clathrin|Sigma-adaptin 1B|Sigma1B-adaptin
- DC22|AP-1 complex subunit sigma-2|Adaptor protein complex AP-1 subunit sigma-1B|Adaptor-related protein complex 1 subunit sigma-1B|Clathrin assembly protein complex 1 sigma-1B small chain|Golgi adaptor HA1/AP1 adaptin sigma-1B subunit|Sigma 1B subunit of AP-1 clathrin|Sigma-adaptin 1B|Sigma1B-adaptin
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Source:
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Animal-free:
- N
- N
- Biological activity:
- Standard
- Standard
- Purity:
- >85%
- >85%
- Concentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Form:
- Liquid
- Liquid
- Formulation:
- pH: 8; Constituents: 40% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0308% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- pH: 8; Constituents: 40% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0308% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- Storage conditions:
- Store at −20 °C
- Store at −20 °C
- Shipping temperature:
- 20
- 20
- Tested applications:
- MS, SDS-PAGE
- MS, SDS-PAGE
Product Family Options
- Marketsource
Specifications
Cat. No.ABCAAB108122-50Protein/peptide nameAP1S2Protein synonymsDC22|AP-1 complex subunit sigma-2|Adaptor protein complex AP-1 subunit sigma-1B|Adaptor-related protein complex 1 subunit sigma-1B|Clathrin assembly protein complex 1 sigma-1B small chain|Golgi adaptor HA1/AP1 adaptin sigma-1B subunit|Sigma 1B subunit of AP-1 clathrin|Sigma-adaptin 1B|Sigma1B-adaptinProtein/peptide typeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedAnimal-freeNBiological activityStandardPurity>85%Concentration0,5 mg/mlFormLiquidFormulationpH: 8; Constituents: 40% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0308% (R*,R*)-1,4-Dimercaptobutan-2,3-diolStorage conditionsStore at −20 °CShipping temperature20Tested applicationsMS, SDS-PAGEMarketsource - Marketsource
Specifications
Cat. No.ABCAAB108122-250Protein/peptide nameAP1S2Protein synonymsDC22|AP-1 complex subunit sigma-2|Adaptor protein complex AP-1 subunit sigma-1B|Adaptor-related protein complex 1 subunit sigma-1B|Clathrin assembly protein complex 1 sigma-1B small chain|Golgi adaptor HA1/AP1 adaptin sigma-1B subunit|Sigma 1B subunit of AP-1 clathrin|Sigma-adaptin 1B|Sigma1B-adaptinProtein/peptide typeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedAnimal-freeNBiological activityStandardPurity>85%Concentration0,5 mg/mlFormLiquidFormulationpH: 8; Constituents: 40% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0308% (R*,R*)-1,4-Dimercaptobutan-2,3-diolStorage conditionsStore at −20 °CShipping temperature20Tested applicationsMS, SDS-PAGEMarketsource



