Auftragserfassung
Germany
ContactUsLinkComponent
Human Complement C2 ELISA Kit
Human Complement C2 ELISA Kit
Catalog # ABCAAB154132-96
Supplier:  Abcam
CAS Number:  
undefined
Human Complement C2 ELISA Kit
Catalog # ABCAAB154132-96
Supplier:  Abcam
CAS Number:  
About this item
Human Complement C2 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Complement C2 in Human in Cerebral Spinal Fluid, plasma, milk, serum samples.