Human Superoxide Dismutase 4/CCS ELISA kit
ABCAAB283972-1X96
: Abcam
:
- Description:Human Superoxide Dismutase 4/CCS ELISA kit
Human Superoxide Dismutase 4/CCS ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Superoxide Dismutase 4/CCS in Human in Saliva, plasma, serum, cell Lysate, Tissue samples.